missing translation for 'onlineSavingsMsg'
Learn More

Levomefolate calcium, MedChemExpress

Product Code. 16403337
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16403337

Brand: Medchem Express HY17383

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Levomefolate calcium is an artificial form of folate.

Levomefolate calcium is an artificial form of folate.

Specifications

Chemical Name or Material Levomefolate calcium
Physical Form Powder
CAS 151533-22-1
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H23CaN7O6
For Use With (Application) Cancer-programmed cell death
Solubility Information H2O : 4.4 mg/mL (8.84 mM; Need ultrasonic and warming)
SMILES O=C([O-])CC[C@@H](C([O-])=O)NC(C1=CC=C(NC[C@@H]2N(C)C3=C(N=C(N)NC3=O)NC2)C=C1)=O.[Ca+2]
Molecular Weight (g/mol) 497.52
Formula Weight 497.52
Percent Purity 97.11%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.