missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lewis Y Antibody (LWY/1463), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54560JF646
This item is not returnable.
View return policy
Description
Lewis Y Monoclonal antibody specifically detects Lewis Y in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Lewis Y | |
| Monoclonal | |
| Janelia Fluor 646 | |
| IPCA-2, IPCA6, IPCA-6, IPCA-8, PCANAP2, PCANAP6, PCANAP8, Prostate cancer-associated protein 6, PRST, Solute carrier family 45 member 3 | |
| Human colon carcinoma cells | |
| 0.1 mL | |
| Cancer, Tumor Biomarkers | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LWY/1463 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction