missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LGALS3 Monoclonal Antibody (OTI8E3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804699
This item is not returnable.
View return policy
Description
Galactose-specific lectin which binds IgE. May mediate with the alpha-3, beta-1 integrin the stimulation by CSPG4 of endothelial cells migration. Together with DMBT1, required for terminal differentiation of columnar epithelial cells during early embryogenesis. In the nucleus: acts as a pre-mRNA splicing factor. Involved in acute inflammatory responses including neutrophil activation and adhesion, chemoattraction of monocytes macrophages, opsonization of apoptotic neutrophils, and activation of mast cells.Specifications
| LGALS3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P17931 | |
| LGALS3 | |
| Full length human recombit protein of human LGALS3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI8E3 | |
| Unconjugated | |
| LGALS3 | |
| CBP35, GAL3, GALBP, GALIG, L31, LGALS2, MAC2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3958 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction