missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LGALS3BP Recombinant Rabbit Monoclonal Antibody (K01_1T47)

Product Code. p-200114075
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30825389 100 μL
30825925 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30825389 Supplier Invitrogen™ Supplier No. MA563496

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

LGALS3BP Recombinant Monoclonal Antibody for Western Blot

The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. LGALS3BP has been found elevated in the serum of patients with cancer and in those infected by the human immunodeficiency virus (HIV). It appears to be implicated in immune response associated with natural killer (NK) and lymphokine-activated killer (LAK) cell cytotoxicity. Using fluorescence in situ hybridization the full length 90K cDNA has been localized to chromosome 17q25. The native protein binds specifically to a human macrophage-associated lectin known as Mac-2 and also binds galectin 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LGALS3BP
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1T47
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene LGALS3BP
Gene Accession No. Q08380
Gene Alias 90K; basement membrane autoantigen p105; BTBD17B; CyCAP; Cyp-C-associated protein; galectin 3 binding protein; galectin-3-binding protein; gp90; L3 antigen; Lectin; lectin galactoside-binding soluble 3 binding protein; lectin galactoside-binding soluble 3-binding protein; lectin, galactoside binding soluble 3 binding protein; lectin, galactoside-binding, soluble, 3 binding protein; Lgals3bp; M2BP; mac-2 BP; mac-2-binding protein; MAC2BP; MAC-2BP; MAC-2-BP; mama; peptidylprolyl isomerase C-associated protein; Ppicap; Protein 90K; Protein MAMA; serum protein 90K; similar to Galectin-3 binding protein precursor (Lectin galactoside-binding soluble 3 binding protein) (Mac-2 binding protein) (Mac-2 BP) (MAC2BP) (Tumor-associated antigen 90K); TANGO10B; transport and golgi organization 10 homolog B; tumor-associated antigen 90K
Gene Symbols LGALS3BP
Host Species Rabbit
Immunogen Recombinant protein of human LGALS3BP.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3959
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.