missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LGALS9 Mouse anti-Human, Clone: OTI19H8, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805651S
This item is not returnable.
View return policy
Description
LGALS9 Monoclonal antibody specifically detects LGALS9 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| LGALS9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O00182 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3965 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI19H8 | |
| Unconjugated | |
| Lgals9 | |
| 36 kDa beta-galactoside-binding lectin; AA407335; AI194909; AI265545; ecalectin; Gal9; gal-9; galectin 9; Galectin9; galectin-9; HOM HD 21; HUAT; Lectin galactose binding soluble 9 (Galectin-9); lectin, galactose binding, soluble 5; lectin, galactose binding, soluble 9; Lectin, galactose binding, soluble 9 (Galectin-9); lectin, galactoside binding soluble 9; lectin, galactoside-binding, soluble, 9; LGALS35; Lgals5; LGALS9; LGALS9A; tumor antigen HOM-HD-21; UAT; urate transporter/channel; urate transporter/channel protein | |
| Full length human recombinant protein of human LGALS9 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction