missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lgi1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76379B
This item is not returnable.
View return policy
Description
Lgi1 Polyclonal antibody specifically detects Lgi1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| Lgi1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| Biotin | |
| epilepsy, partial, EPITEMPIN, Epitempin-1, EPTleucine-rich glioma-inactivated protein 1, ETL1epitempin-1, IB1099, leucine-rich, glioma inactivated 1 | |
| Antibody was raised against a 13 amino acid synthetic peptide from near the amino terminus of human LGI1. The immunogen is located within the first 50 amino acids of LGI1. Amino Acid Squence: LLLTEGKKPAKPK | |
| 0.1 mL | |
| Neuroscience | |
| 9211 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction