missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lgi1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77320-0.025mg
This item is not returnable.
View return policy
Description
Lgi1 Polyclonal specifically detects Lgi1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| Lgi1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| epilepsy, partial, EPITEMPIN, Epitempin-1, EPTleucine-rich glioma-inactivated protein 1, ETL1epitempin-1, IB1099, leucine-rich, glioma inactivated 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This LGI1 antibody is predicted to be specific to LGI1 and not recognize other LGI proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| AAQ89244 | |
| LGI1 | |
| Antibody was raised against a 14 amino acid synthetic peptide from near the center of human LGI1. The immunogen is located within amino acids 370 - 420 of LGI1. Amino Acid Squence: LEIVRTPQTLRTPH | |
| 0.025 mg | |
| Neuroscience | |
| 9211 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction