missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lgi1 Antibody (S283-7), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42190JF549
This item is not returnable.
View return policy
Description
Lgi1 Monoclonal antibody specifically detects Lgi1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Lgi1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| epilepsy, partial, EPITEMPIN, Epitempin-1, EPTleucine-rich glioma-inactivated protein 1, ETL1epitempin-1, IB1099, leucine-rich, glioma inactivated 1 | |
| Fusion protein amino acids 37-113 (LRRNT domain and first LRR repeat) of mouse LGI1. Rat: 100% identity (77/77 amino acids identical). Human: 98% identity (76/77 amino acids identical). ∽50% identity with LGI2, LGI3 and LGI4. | |
| 0.1 mL | |
| Neuroscience | |
| 9211 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Immunohistochemistry, Immunofluorescence | |
| S283-7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction