missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lgi1 Antibody (SR2092), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | Lgi1 |
|---|---|
| Clone | SR2092 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18602884
|
Novus Biologicals
NBP3-22331-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18614144
|
Novus Biologicals
NBP3-22331-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lgi1 Monoclonal antibody specifically detects Lgi1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
| Lgi1 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| epilepsy, partial, EPITEMPIN, Epitempin-1, EPTleucine-rich glioma-inactivated protein 1, ETL1epitempin-1, IB1099, leucine-rich, glioma inactivated 1 | |
| A synthesized peptide derived from human Lgi1 (Uniprot #: O95970) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2092 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 9211 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title