missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LGI1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA520506
This item is not returnable.
View return policy
Description
A suggested positive control is mouse brain tissue lysate. PA5-20506 can be used with blocking peptide PEP-0626.
The leucine-rich, glioma inactivated gene 1 (LGI1) was first identified as a candidate tumor suppressor gene for glioma and may play a role in other cancers. LGI1 is a member of a family of highly related proteins containing leucine-rich repeats (LRRs) which are highly similar to other transmembrane signaling molecules and receptors. LGI1 serves as a ligand to ADAM22, a metalloprotease localized at the synapse. Mutations in LGI1 account for nearly half of autodominant lateral temporal epilepsy (ADTLE), an epileptic syndrome characterized by focal seizures with predominant auditory symptoms. Two isoforms of LGI1 are known to exist.
Specifications
| LGI1 | |
| Polyclonal | |
| Unconjugated | |
| LGI1 | |
| ADLTE; ADPAEF; ADPEAF; BB130740; EPITEMPIN; Epitempin 1; Epitempin-1; EPT; ETL1; IB1099; leucine rich glioma inactivated 1; leucine-rich glioma inactivated 1; leucine-rich glioma-inactivated protein 1; leucine-rich repeat LGI family, member 1; leucine-rich, glioma inactivated 1; LGI1; OTTHUMP00000020121; OTTHUMP00000020122; UNQ775/PRO1569 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 252892, 56839, 9211 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O95970, Q8K4Y5, Q9JIA1 | |
| LGI1 | |
| A 13 amino acid peptide from near the amino terminus of human LGI1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction