missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LGMN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112345
This item is not returnable.
View return policy
Description
LGMN Polyclonal Antibody for Western Blot, IP
This gene encodes a cysteine protease that has a strict specificity for hydrolysis of asparaginyl bonds. This enzyme may be involved in the processing of bacterial peptides and endogenous proteins for MHC class II presentation in the lysosomal/endosomal systems. Enzyme activation is triggered by acidic pH and appears to be autocatalytic. Protein expression occurs after monocytes differentiate into dendritic cells. A fully mature, active enzyme is produced following lipopolysaccharide expression in mature dendritic cells. Overexpression of this gene may be associated with the majority of solid tumor types. This gene has a pseudogene on chromosome 13. Several alternatively spliced transcript variants have been described, but the biological validity of only two has been determined.
Specifications
| LGMN | |
| Polyclonal | |
| Unconjugated | |
| LGMN | |
| AEP; AI746452; Asparaginyl endopeptidase; AU022324; cb173; cb460; cysteine protease 1; legumain; Lgmn; LGMN1; preprolegumain; protease, cysteine 1; protease, cysteine, 1; protease, cysteine, 1 (legumain); PRSC1; PRSC1EC 3.4.22.34; sb:cb173; wu:fa98g05; wu:fj92g08; zgc:76953 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100154477 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| 0 | |
| LGMN | |
| Recombinant Sus scrofa (Pig) Legumain/LGMN Protein. | |
| 100 μL | |
| Primary | |
| Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction