Learn More
Invitrogen™ LGMN Recombinant Rabbit Monoclonal Antibody (23GB2095), Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA554040
 View more versions of this product
Product Code. 30088164
  • € 562.00 / 100µL
Description
This gene encodes a cysteine protease that has a strict specificity for hydrolysis of asparaginyl bonds. This enzyme may be involved in the processing of bacterial peptides and endogenous proteins for MHC class II presentation in the lysosomal/endosomal systems. Enzyme activation is triggered by acidic pH and appears to be autocatalytic. Protein expression occurs after monocytes differentiate into dendritic cells. A fully mature, active enzyme is produced following lipopolysaccharide expression in mature dendritic cells. Overexpression of this gene may be associated with the majority of solid tumor types. This gene has a pseudogene on chromosome 13. Several alternatively spliced transcript variants have been described, but the biological validity of only two has been determined.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LGMN
Recombinant Monoclonal
0.8 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q99538, Q9R0J8
LGMN
A synthesized peptide derived from human Legumain (200-350AA).
100 μL
Primary
Human, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB2095
Unconjugated
LGMN
AEP; AI746452; Asparaginyl endopeptidase; AU022324; cb173; cb460; cysteine protease 1; legumain; Lgmn; LGMN1; preprolegumain; protease, cysteine 1; protease, cysteine, 1; protease, cysteine, 1 (legumain); PRSC1; PRSC1EC 3.4.22.34; sb:cb173; wu:fa98g05; wu:fj92g08; zgc:76953
Rabbit
Affinity chromatography
RUO
5641, 63865
-20°C