missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lgr4/GPR48 Antibody (OTI2D1), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72006APC
This item is not returnable.
View return policy
Description
Lgr4/GPR48 Monoclonal specifically detects Lgr4/GPR48 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| Lgr4/GPR48 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| LGR4 | |
| Recombinant protein fragment corresponding to amino acids 230-540 of human Lgr4/GPR48 (NP_060960 ) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI2D1 | |
| Flow Cytometry | |
| GPR48G protein-coupled receptor 48, G-protein coupled receptor 48, leucine-rich repeat containing G protein-coupled receptor 4 | |
| Mouse | |
| Affinity Purified | |
| GPCR | |
| 55366.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction