missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LGR4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17780224
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17780224 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17780224 Supplier Invitrogen™ Supplier No. PA5118812

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR48.

G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR LGR4 is a member of this family (subfamily Orphan-A). Its expression has been documented in human brain, adrenal, heart, kidney, liver, skeletal muscle, ovary, pancreas, placenta, prostate, spleen, stomach, testis, thyroid, and spinal cord.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LGR4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LGR4
Gene Accession No. A2ARI4, Q9BXB1, Q9Z2H4
Gene Alias 9130225G07; A330106J01Rik; A930009A08Rik; BNMD17; G protein-coupled receptor 48; G protein-coupled receptor GPR48; GPR48; G-protein coupled receptor 48; leucine rich repeat containing G protein-coupled receptor 4; leucine-rich repeat containing G protein-coupled receptor 4; leucine-rich repeat-containing G protein-coupled receptor 4; leucine-rich repeat-containing G-protein coupled receptor 4; Lgr4
Gene Symbols LGR4
Host Species Rabbit
Immunogen A synthesized peptide derived from human LGR4(Accession Q9BXB1), corresponding to amino acid residues L884-S934.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107515, 286994, 55366
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.