missing translation for 'onlineSavingsMsg'
Learn More

Lgr5/GPR49 Antibody (CL13579) [DyLight 594], Novus Biologicals™

Product Code. 30523579 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30523579 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30523579 Supplier Novus Biologicals Supplier No. NBP343804DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lgr5/GPR49 Monoclonal antibody specifically detects Lgr5/GPR49 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lgr5/GPR49
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL13579
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias FEX, G protein-coupled receptor 49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of O75473, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Neuroscience, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8549
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.