missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LGR5 Mouse anti-Human, Clone:2B5B9, Abnova™
Mouse monoclonal antibody raised against recombinant human LGR5.
Brand: Abnova MAB17138.100ug
This item is not returnable.
View return policy
Specifications
| LGR5 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| LGR5 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Flow Cytometry, Western Blot | |
| 2B5B9 | |
| leucine-rich repeat-containing G protein-coupled receptor 5 | |
| In PBS (0.05% sodium azide) | |
| FEX, GPR49, GPR67, GRP49, HG38, MGC117008 | |
| Recombinant protein corresponding to amino acid 22-178 of human LGR5 from E. coli. | |
| 8549 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction