missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lgr6 Antibody (OTI2A3), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72119AF750
This item is not returnable.
View return policy
Description
Lgr6 Monoclonal specifically detects Lgr6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Lgr6 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| LGR6 | |
| Recombinant protein fragment corresponding to amino acids 25-250 of human Lgr6 (NP_001017403) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI2A3 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| FLJ14471, gonadotropin receptor, GPCR, leucine-rich repeat containing G protein-coupled receptor 6, leucine-rich repeat-containing G protein-coupled receptor 6, leucine-rich repeat-containing G-protein coupled receptor 6, VTS20631 | |
| Mouse | |
| Affinity Purified | |
| GPCR | |
| 59352.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction