missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LGR6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17117843
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17117843 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17117843 Supplier Invitrogen™ Supplier No. PA5109909

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: QLPRLRVLEL SHNQIEELPS LHRCQKLEEI GLQHNRIWEI GADTFSQLSS LQALDLSWNA IRSIHPEAFS TLHSLVKLDL TDNQLTTLPL AGLGGLMHLK LKGNLALSQA FSKDSFPKLR ILEVPYAYQC CPYGMCASFF KASGQWEAED LHLDDEESSK R.

LGR6 has been reported to be expressed in rat adrenal, brain, colon, heart, kidney, ovary, oviduct, small intestine, spleen, testis, and uterus. ESTs have been isolated from human B-cell/lung/testis, blood, brain, colon, eye, germ cell, head/neck, heart, heart/melanocyte/uterus, lung, ovary, stomach, testis, and uterus libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LGR6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Lgr6
Gene Accession No. Q3UVD5, Q9HBX8
Gene Alias A530037C04Rik; D830026M09; FLJ14471; gonadotropin receptor; GPCR; leucine rich repeat containing G protein-coupled receptor 6; leucine-rich repeat containing G protein-coupled receptor 6; leucine-rich repeat-containing G protein-coupled receptor 6; leucine-rich repeat-containing G-protein coupled receptor 6; Lgr6; RGD1560339; RP11-572A16.3; UNQ6427/PRO21331; VTS20631
Gene Symbols Lgr6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 350-510 of human LGR6 (NP_0010174031).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 329252, 498233, 59352
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.