missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LGSN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00 - € 624.00
Specifications
| Antigen | LGSN |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18230384
|
Novus Biologicals
NBP2-58511 |
100 μL |
€ 624.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18678887
|
Novus Biologicals
NBP2-58511-25ul |
25 μL |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LGSN Polyclonal specifically detects LGSN in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| LGSN | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 51557 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:VSRSKTIPAHFFQEKVSHGVCMPRGYLEVIPNPKDNEMNNIRATCFNSDIVLMPELSTFRVLPWADRTARVICDTFTVTGEPLLTSPRY | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| GLULD1, glutamate-ammonia ligase (glutamine synthase) domain containing 1, glutamate-ammonia ligase (glutamine synthetase) domain containing 1, Glutamate-ammonia ligase domain-containing protein 1, lengsin, lengsin, lens protein with glutamine synthetase domain, Lens glutamine synthase-like, LGSMGC163240, MGC163238 | |
| LGSN | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title