missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHR Antibody (LHCGR/1416), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54487APCCY7
This item is not returnable.
View return policy
Description
LHR Monoclonal specifically detects LHR in Human samples. It is validated for Flow Cytometry.
Specifications
| LHR | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| LHCGR | |
| Recombinant fragment (around aa 70-410) of human LHR protein (exact sequence is proprietary) (Uniprot: P22888) | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| LHCGR/1416 | |
| Flow Cytometry | |
| FLJ41504, HHG, hypergonadotropic hypogonadism, LCGRLH/CGR, LH/CG-R, LHRHR, LHRLGR2ULG5, LSH-R, Luteinizing hormone receptor, luteinizing hormone/choriogonadotropin receptor, lutropin/choriogonadotropin receptor, lutropin-choriogonadotropic hormone receptor | |
| Mouse | |
| Protein A or G purified | |
| GPCR | |
| 3973.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction