missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LHR Antibody (LHCGR/7399) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26966IR
This item is not returnable.
View return policy
Description
LHR Monoclonal antibody specifically detects LHR in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| LHR | |
| Monoclonal | |
| DyLight 755 | |
| FLJ41504, HHG, hypergonadotropic hypogonadism, LCGRLH/CGR, LH/CG-R, LHRHR, LHRLGR2ULG5, LSH-R, Luteinizing hormone receptor, luteinizing hormone/choriogonadotropin receptor, lutropin/choriogonadotropin receptor, lutropin-choriogonadotropic hormone receptor | |
| Recombinant fragment (around aa 100-300) of human LHR protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Caveolae Markers, GPCR | |
| 3973 | |
| Store at 4°C in the dark. | |
| IgG2 κ |
| Immunohistochemistry (Paraffin) | |
| LHCGR/7399 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction