Learn More
Invitrogen™ LHX2 Monoclonal Antibody (OTI2E8)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA527283
Product Code. 15946874
  • € 570.00 / 100µL
Description
LHX2 Monoclonal Antibody for Western Blot
This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator. The protein can recapitulate or rescue phenotypes in Drosophila caused by a related protein, suggesting conservation of function during evolution.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LHX2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P50458, Q9Z0S2
Lhx2
Human recombinant protein fragment corresponding to amino acids 220-406 of LHX2 produced in E.coli.
100 μL
Primary
Human, Mouse
Antibody
IgG2b
Western Blot
OTI2E8
Unconjugated
Lhx2
ap; apterous; hLhx2; Homeobox protein LH-2; LH2; Lh-2; LH2A; LHX2; LIM homeo box protein 2; LIM homeobox 2; LIM homeobox protein 2; LIM HOX gene 2; LIM/homeobox protein Lhx2; Lim2; MGC138390
Mouse
Affinity chromatography
RUO
16870, 9355
-20°C, Avoid Freeze/Thaw Cycles