missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHX2 Mouse anti-Human, Clone: OTI2H2, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810303
This item is not returnable.
View return policy
Description
This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator. The protein can recapitulate or rescue phenotypes in Drosophila caused by a related protein, suggesting conservation of function during evolution.Specifications
| LHX2 | |
| Monoclonal | |
| Unconjugated | |
| LHX2 | |
| LH2, hLhx2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9355 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI2H2 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P50458 | |
| LHX2 | |
| Human recombit protein fragment corresponding to amino acids 220-406 of human LHX2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG3 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction