missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LHX3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103073
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total LHX3.
This gene encodes a member a large protein family which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein is a transcription factor that is required for pituitary development and motor neuron specification. Mutations in this gene have been associated with a syndrome of combined pituitary hormone deficiency and rigid cervical spine. Two transcripts variants encoding distinct isoforms have been identified for this gene.
Specifications
| LHX3 | |
| Polyclonal | |
| Unconjugated | |
| LHX3 | |
| bLhx3b; CPHD3; DKFZp762A2013; Homeobox protein LIM-3; homeobox protein P-LIM; lhx3; Lhx3b; LIM homeo box protein 3; LIM homeobox 3; LIM homeobox protein; LIM homeobox protein 3; LIM homeodomain 3; LIM homeodomain 3 protein b isoform; LIM homeodomain box protein 3; LIM homeodomain protein 3b; LIM/homeobox protein Lhx3; LIM/homeodomain protein LHX3; lim3; Lim-3; LOW QUALITY PROTEIN: LIM/homeobox protein Lhx3; M2-LHX3; mLIM3; mLim-3; Plim; P-LIM | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16871, 170671, 8022 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P50481, Q9UBR4 | |
| LHX3 | |
| A synthesized peptide derived from human LHX3(Accession Q9UBR4), corresponding to amino acid residues P357-F397. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction