missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LHX5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540683
This item is not returnable.
View return policy
Description
Peptide sequence: FFRSPRRMRP LGGRLDESEM LGSTPYTYYG DYQGDYYAPG SNYDFFAHGP Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 83%.
This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and be involved in the control of differentiation and development of the forebrain. In mice, this protein is essential for the regulation of precursor cell proliferation and the control of neuronal differentiation and migration during hippocampal development. This protein is involved in learning and motor functions in adult mice.
Specifications
| LHX5 | |
| Polyclonal | |
| Unconjugated | |
| LHX5 | |
| homeobox protein LIM-2; Homeobox protein LIM-5; Lhx5; LIM class homeodomain protein; LIM homeo box protein 5; LIM homeobox 5; LIM homeobox protein 5; LIM/homeobox protein Lhx5; Lim2; Lim-2; lim5; LOW QUALITY PROTEIN: LIM/homeobox protein Lhx5; MGC129689 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 16873, 64211 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P61375, Q9H2C1 | |
| LHX5 | |
| Synthetic peptide directed towards the middle region of human LHX5. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction