missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHX6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38418-20ul
This item is not returnable.
View return policy
Description
LHX6 Polyclonal antibody specifically detects LHX6 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| LHX6 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| LHX6.1LIM homeobox protein 6, LIM homeobox 6, LIM homeodomain protein 6.1, LIM/homeobox protein Lhx6, LIM/homeobox protein Lhx6.1, MGC119542, MGC119544, MGC119545 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 143-392 of human LHX6 (NP_055183.2).,, Sequence:, NKEIFCKMDYFSRFGTKCARCGRQIYASDWVRRARGNAYHLACFACFSCKRQLSTGEEFGLVEEKVLCRIHYDTMIENLKRAAENGNGLTLEGAVPSEQDSQPKPAKRARTSFTAEQLQVMQAQFAQDNNPDAQTLQKLADMTGLSRRVIQVWFQNCRARHKKHTPQHPVPPSGAPPSRLPSALSDDIHYTPFSSPERARMVTLHGYIESQVQCGQVHCRLPYTAPPVHLKADMDGPLSNRGEKVILFQY | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 26468 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?