Learn More
LHX8 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30722-1-AP-150UL
 View more versions of this product
Product Code. 18025015
  • € 410.00 / 150µL
Description
The LHX8 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human LHX8. This antibody recognizes human antigen. The LHX8 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
LHX8
400 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
Lhx7, LHX8, LIM homeobox 8, LIM homeobox protein 8, LIM/homeobox protein Lhx8
150 μL
431707
Store at -20°C. Stable for one year after shipment.