missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHX9 Antibody (CL15033) [Unconjugated], Novus Biologicals™
Mouse Monoclonal Antibody
€ 422.00 - € 605.00
Specifications
| Antigen | LHX9 |
|---|---|
| Clone | CL15033 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279530
|
Novus Biologicals
NBP3-43644-100ul |
100 μL |
€ 605.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279611
|
Novus Biologicals
NBP3-43644-25ul |
25 μL |
€ 422.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LHX9 Monoclonal antibody specifically detects LHX9 in Human samples. It is validated for Western BlotSpecifications
| LHX9 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cell Biology | |
| PBS, pH 7.2, 40% glycerol | |
| 56956 | |
| IgG2a | |
| Protein A purified |
| CL15033 | |
| Western Blot 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| LIM homeobox 9, LIM homeobox protein 9, LIM/homeobox protein Lhx9 | |
| This antibody was generated using a synthetic peptide of Q9NQ69, with the exact immunogen sequence remaining proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title