missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIAS Polyclonal Antibody
GREENER_CHOICE

Product Code. 17441693 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17441693 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17441693 Supplier Invitrogen™ Supplier No. PA5118032

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LIAS Polyclonal Antibody for Western Blot, IHC (P), IP

LIAS belongs to the biotin and lipoic acid synthetases family. It localizes in mitochondrion and plays an important role in alpha-(+)-lipoic acid synthesis. It may also function in the sulfur insertion chemistry in lipoate biosynthesis.The protein encoded by this gene belongs to the biotin and lipoic acid synthetases family. It localizes in mitochondrion and plays an important role in alpha-(+)-lipoic acid synthesis. It may also function in the sulfur insertion chemistry in lipoate biosynthesis. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.The protein encoded by this gene belongs to the biotin and lipoic acid synthetases family. It localizes in mitochondrion and plays an important role in alpha-(+)-lipoic acid synthesis. It may also function in the sulfur insertion chemistry in lipoate biosynthesis. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIAS
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene Lias
Gene Accession No. O43766
Gene Alias 2900022L22Rik; 4933425M12Rik; 7a5ex; C77512; HAMAP-Rule:MF_03123}; HGCLAS; HUSSY-01; hypothetical protein MGC66080; LAS; LIAS; LIAS protein; LIP1; Lipoate synthase; lipoate synthase {ECO:0000255; lipoic acid synthase; lipoic acid synthase {ECO:0000255; lipoic acid synthetase; lipoic acid synthetase isoform 1 precursor; lipoyl synthase, mitochondrial; lipoyl synthase, mitochondrial {ECO:0000255; Lipoyl synthase, mitochondrial-like protein; lip-syn; lip-syn {ECO:0000255; LS; LS {ECO:0000255; MGC23245; mLip1; PDHLD; zgc:66080
Gene Symbols Lias
Host Species Rabbit
Immunogen E. coli-derived Human LIAS fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11019
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.