missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIF Antibody (39N7D10), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-27406H
This item is not returnable.
View return policy
Description
LIF Monoclonal specifically detects LIF in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| LIF | |
| Monoclonal | |
| HRP | |
| CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 39N7D10 | |
| Western Blot, Immunohistochemistry-Paraffin | |
| LIF | |
| A recombinant murine Lif protein containing amino acids 24-203 was used as the immunogen for this antibody. | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Lines | |
| 3976 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction