Learn More
LIF Antibody (9808), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB2501T
Product Code. 30059594
  • € 865.00 / 0.50mL
Description
LIF Monoclonal antibody specifically detects LIF in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LIF
Monoclonal
Alexa Fluor 594
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, CyTOF
9808
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI
E. coli-derived recombinant human LIF
0.5 mL
Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Lines
3976
Store at 4C in the dark.