missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIF Antibody (9808), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2501E
This item is not returnable.
View return policy
Description
LIF Monoclonal antibody specifically detects LIF in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| LIF | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 9808 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI | |
| E. coli-derived recombinant human LIF | |
| 0.5 mL | |
| Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Lines | |
| 3976 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction