missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIF Antibody (MM0454-6H31), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11773MFV450
This item is not returnable.
View return policy
Description
LIF Monoclonal antibody specifically detects LIF in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization
Specifikationer
| LIF | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization | |
| MM0454-6H31 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize | |
| CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI | |
| Human recombinant LIF | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Lines | |
| 3976 | |
| Store at 4C in the dark. | |
| IgG2 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering