Learn More
Invitrogen™ E-Gel™ Imager Quantitation USB Dongle
Description
Includes
1 x Gel Quant Express software enabling USB Dongle
To use this powerful quantitation and analysis software, insert an activation dongle into a free USB port on PC whenever the software is in use. With additional USB dongles, software can be used on more than one computer simultaneously.
- Access powerful Gel Quant Express software using USB dongle
- Key to Quantitation Gel Quant Express is powerful and intuitive software tool included with each E-Gel Imager system
- In order to utilize software, HASP key in form of USB dongle must be inserted into free USB port on computer
- Each E-Gel Imager system includes single Quantitation Software USB Dongle that will allow single instance of Gel quant Express software to be used at given time
- Separate USB dongle is required for each instance of Gel Quant Express Software that will be used at given time
- Compact Solution for Agarose Gel Imaging, Analysis, and Documentation
- Powerful and flexible system that allows to document gel images using multiple nucleic acid dyes and even Qdottrademark (TM) nanocrystals
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Specifications
Specifications
| Content And Storage | 1 x Gel Quant Express software enabling USB Dongle. Store at Room temperature. |
| For Use With (Equipment) | E-Gel Imager |
| Product Line | E-Gel |
| Type | Imager Quantitation USB Dongle |
| Operating System | Windows XP (Service Pack 3), Windows 7 |
| Shipping Condition | Room Temperature |
| Software Category | Validation and Verification Software |
| Software Type | Image Analysis |
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.