missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIFR alpha Antibody (1), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89586CL1
This item is not returnable.
View return policy
Description
LIFR alpha Monoclonal antibody specifically detects LIFR alpha in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| LIFR alpha | |
| Monoclonal | |
| CoraFluor 1 | |
| CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF receptor, LIF-R, SJS2, STWS, SWS | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human LIFR alpha (Accession#: NP_001121143.1; Met 1-Ser 833). | |
| 0.1 mL | |
| Immunology | |
| 3977 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| 1 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction