missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIFR alpha Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32070IR
This item is not returnable.
View return policy
Description
LIFR alpha Polyclonal antibody specifically detects LIFR alpha in Human, Mouse samples. It is validated for Western Blot
Specifications
| LIFR alpha | |
| Polyclonal | |
| Western Blot | |
| CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF receptor, LIF-R, SJS2, STWS, SWS | |
| The immunogen this antibody was made to, maps to a region between residue 1047 to 1097 of human Leukemia Inhibitory Factor Receptor Alpha using the numbering given in entry NP_001121143.1 (GeneID 3977). | |
| 0.1 mL | |
| Immunology | |
| 3977 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction