missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIFR alpha Antibody (MM0455-9B23), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11774APCCY7
This item is not returnable.
View return policy
Description
LIFR alpha Monoclonal specifically detects LIFR alpha in Human samples. It is validated for Flow Cytometry.
Specifications
| LIFR alpha | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| LIFR | |
| Human recombinant LIFR alpha (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| MM0455-9B23 | |
| Flow Cytometry | |
| CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF receptor, LIF-R, SJS2, STWS, SWS | |
| Mouse | |
| Protein G purified | |
| Immunology | |
| 3977.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction