missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIG3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26583-1-AP-150UL
This item is not returnable.
View return policy
Description
There are four distinct ligase proteins found in mammalian cells. The activity of these DNA ligases is required for the joining of DNA strand breaks that occur during DNA replication, repair and recombination. DNA Ligase III gene product migrates as a 100 kDa (a-form) protein in somatic tissues, is ubiquitously expressed and complexes with XRCC-1 and a 97 kDa (beta form) protein that is found in testis. DNA Ligase III is involved in rejoining DNA single-strand breaks and in the base excision repair pathway.Specifications
| LIG3 | |
| Polyclonal | |
| Unconjugated | |
| LIG3 | |
| DNA ligase 3, DNA ligase III, LIG2, LIG3, ligase III, DNA, ATP dependent | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16882, 3980 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P49916, P97386 | |
| LIG3 | |
| LIG3 Fusion Protein Ag24998 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction