Learn More
Abeba™ Light 1038 Shoes
Product Code. 15689914
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15689914
Brand: Abeba™ 1038/35
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
  • CE EN ISO 20345:2011 S2 SRA
  • Smooth leather
  • Black
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3557)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S2 SRA
Color Black
Material Leather
Size 35
Sole Features Slip-resistant
Sole Material Polyurethane
Style 1038
Toe Material Steel