Learn More
Memmert™ Light Module Warm White 2,700 K
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP110eco
HPP260eco
HPP410eco
HPP750eco
Unit Size:
Each
Description
- 10 strips for model 110
- 14 for model 260/410/750
- Programme-controlled dimming rom 0 to 100% (in 1 % steps)
- Ramp programming in combination with temperature - only with TwinDISPLAY
- Not in combination with entry ports F6, F7, D8
Specifications
Specifications
| Color | White |
| For Use With (Equipment) | HPP260eco |