Learn More
LIGHT/TNFSF14 Antibody (906909), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB17943H
Product Code. 18615273
  • € 608.00 / 0.10mL
Description
LIGHT/TNFSF14 Monoclonal antibody specifically detects LIGHT/TNFSF14 in Mouse samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LIGHT/TNFSF14
Monoclonal
HRP
PBS
Rat
0.1 mL
Apoptosis
8740
Store at 4C in the dark.
Flow Cytometry, CyTOF
906909
Flow Cytometry, CyTOF-ready
CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT
Mouse myeloma cell line NS0-derived recombinant mouse LIGHT/TNFSF14, Asp72-Val239, Accession # Q9QYH9