Learn More
LIGHT/TNFSF14 Antibody (MM0456-8C34), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-11964G
Product Code. 18058649
  • ONLINE EXCLUSIVE PRICE
    € 642.00
    € 606.90 / 0.10mL
    Save € 35.10
    5% Off
Description
LIGHT/TNFSF14 Monoclonal specifically detects LIGHT/TNFSF14 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LIGHT/TNFSF14
Monoclonal
DyLight 488
O43557
TNFSF14
Human recombinant LIGHT/TNFSF14 (extracellular domain)
0.1 mL
Apoptosis
8740
Store at 4C in the dark.
IgG1
Flow Cytometry
MM0456-8C34
Flow Cytometry
CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT
Mouse
Protein G purified