missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIGHT/TNFSF14 Antibody (MM0456-8C34), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11964DL594
This item is not returnable.
View return policy
Description
LIGHT/TNFSF14 Monoclonal antibody specifically detects LIGHT/TNFSF14 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| LIGHT/TNFSF14 | |
| Monoclonal | |
| DyLight 594 | |
| CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT | |
| Human recombinant LIGHT/TNFSF14 (extracellular domain) | |
| 0.1 mL | |
| Apoptosis | |
| 8740 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| MM0456-8C34 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction