Learn More
Novus Biologicals™ Lightning-Link (R) Rapid DyLight 594 Antibody Labeling Kit
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
Lightning-Link™ Rapid Antibody Labeling kits enable direct labeling of antibodies, proteins, peptides or other biomolecules and allow conjugations to be set up in seconds, and used within minutes (less than 20 minutes).
Kit Contents:
- Lightning-Link™ Rapid mix
- 1 vial of LL Rapid Quencher reagent
- 1 vial of LL Rapid Modifier reagent
Specifications
Specifications
| Quantity | 100 μg |
| Product Type | Antibody Labeling Kit |
| Content And Storage | -20°C |