missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid Rhodamine Antibody Labeling Kit

Product Code. 18053674 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18053674

Brand: Novus Biologicals™ 3110005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody labeling kit

Lightning-Link™ Rapid Antibody Labeling kits enable direct labeling of antibodies, proteins, peptides or other biomolecules and allow conjugations to be set up in seconds, and used within minutes (less than 20 minutes).

Kit Contents:

  • Lightning-Link™ Rapid mix
  • 1 vial of LL Rapid Quencher reagent
  • 1 vial of LL Rapid Modifier reagent
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 μg
Product Type Antibody Labeling Kit
Content And Storage -20°C
Target Lightning-Link (R) Rapid Rhodamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.