missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LILRA1/LILRB1 Antibody (586326), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB30851PECY55
This item is not returnable.
View return policy
Description
LILRA1/LILRB1 Monoclonal antibody specifically detects LILRA1/LILRB1 in Human samples. It is validated for Flow Cytometry
Specifications
| LILRA1/LILRB1 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11024 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry | |
| 586326 | |
| Flow Cytometry | |
| CD85I, LIR6, LIR-6 | |
| Mouse myeloma cell line NS0-derived recombinant human LILRA1, Pro17-Asn461, Accession # O75019 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction