missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LILRA2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17147843
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17147843 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17147843 Supplier Invitrogen™ Supplier No. PA5109912

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GHLPKPTLWA EPGSVIIQGS PVTLRCQGSL QAEEYHLYRE NKSASWVRRI QEPGKNGQFP IPSITWEHAG RYHCQYYSHN HSSEYSDPLE LVVTGAYSKP TLSALPSPVV TSGGNVTLQC VSQVAFDGFI LCKEGEDEHP QRLNSHSHAR GWSWAIFSVG PVSPSRRWSY RCYAYDSNSP YVWSLPSDLL ELLVPGVSKK PSLSVQPGPM VAPGESL.

Leukocyte Ig-like receptors (LIRs) are a family of immunoreceptors expressed predominantly on monocytes and B cells and at lower levels on dendritic cells and natural killer (NK) cells. All LIRs in subfamily B have an inhibitory function (LILRB1; MIM 604811). LIRs in subfamily A, with short cytoplasmic domains lacking an immunoreceptor tyrosine-based inhibitory motif (ITIM) and with trans membrane regions containing a charged arginine residue, may initiate stimulatory cascades. One member of subfamily A (LILRA3; MIM 604818) lacks a transmembrane region and is presumed to be a soluble receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LILRA2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LILRA2
Gene Accession No. Q8N149
Gene Alias CD85 antigen-like family member H; CD85h; ILT1; ILT-1; Immunoglobulin-like transcript 1; leucocyte Ig-like receptor A2; leukocyte immunoglobulin like receptor A2; leukocyte immunoglobulin-like receptor 7; Leukocyte immunoglobulin-like receptor subfamily A member 2; leukocyte immunoglobulin-like receptor subfamily A member 2 soluble; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 2; LILRA2; LIR7; LIR-7
Gene Symbols LILRA2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 24-240 of human LILRA2 (NP_006857.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11027
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.