missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LILRB3 Mouse anti-Human, Clone: OTI3G9, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812163S
This item is not returnable.
View return policy
Description
LILRB3 Monoclonal antibody specifically detects LILRB3 in Human samples. It is validated for Western BlotSpecifications
| LILRB3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75022 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11025 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI3G9 | |
| Unconjugated | |
| LILRB3 | |
| CD85 antigen-like family member A; CD85A; Cell-surface glycoprotein p91; Gp91; HL9; ILT5; ILT-5; immunoglobulin-like transcript 5; leucocyte Ig-like receptor B3; leukocyte immunoglobulin like receptor B3; leukocyte immunoglobulin-like receptor 3; leukocyte immunoglobulin-like receptor subfamily B member 3; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 6; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 3; LILRA6; Lilrb3; LIR3; LIR-3; monocyte inhibitory receptor HL9; paired Ig-like receptor B; paired immunoglobulin-like receptor B; PIRB; PIR-B | |
| Human recombinant protein fragment corresponding to amino acids 465-631 of human LILRB3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction