missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIM Kinase 1/2 [p Thr508] Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54645CL1
This item is not returnable.
View return policy
Description
LIM Kinase 1/2 Polyclonal antibody specifically detects LIM Kinase 1/2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| LIM Kinase 1/2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Epitope affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| EC 2.7.11.1, LIM domain kinase 1, LIM motif-containing protein kinase, LIMKLIMK-1 | |
| A peptide sequence around phosphorylation site of threonine 508 (R-Y-T (p) -V-V) derived from Human LIMK1 | |
| 0.1 mL | |
| Breast Cancer, Cancer, Phospho Specific, Protein Kinase | |
| 3984 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction