missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIM Kinase 1 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97870V
This item is not returnable.
View return policy
Description
LIM Kinase 1 Polyclonal specifically detects LIM Kinase 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| LIM Kinase 1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 2.7.11.1, LIM domain kinase 1, LIM motif-containing protein kinase, LIMKLIMK-1 | |
| Produced in rabbits immunized with E. coli-derived Human LIM Kinase 1 fragment. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Phospho Specific, Protein Kinase | |
| 3984 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction